Proto® Alloy Steel Socket Driver, Testequity LLC TE
Supplier: TESTEQUITY LLC TE
J5214
76245-200EA 32.11 USD
76245-200
Proto® Alloy Steel Socket Driver, Testequity LLC TE
Tool Sets
This Proto® 12-point socket is ideal for use on stubborn or damaged fasteners, and also on fasteners made from soft materials.
The TorquePlus™ drive configuration allows for a quick and easy socket engagement to the fastener flats, not the corners.
Features with a radius-corner design that shifts contact area and stress away from the fastener corners and distributes contact stresses over a large area of the socket, minimizing slippage. The internal design minimizes fastener rounding and allows the user to more easily turn worn, rounded, or rusted fasteners.
Certifications: Meets ASME B107.1 and AS954E S3.8.1 specifications.