Thermally Conductive Tapes, 3M
Supplier: 3M
7010311896
76245-048CS 3756.88 USD
76245-048
Thermally Conductive Tapes, 3M
Tapes
Thermally Conductive Adhesive Transfer Tapes are designed to provide a preferential heat-transfer path between heat-generating components and heat sinks or other cooling devices.
  • High mechanical strength
  • Improved surface wet-out for rough surface
  • Excellent shock performance
  • Wider and longer roll is available
  • Halogen free and good thermal transfer
  • Ideal for thin bonding applications
These tapes are pressure-sensitive adhesives loaded with thermally conductive ceramic fillers. Pressure is needed to form a bond. Heat and pressure may be needed on some substrates to achieve an acceptable bond. The specialized acrylic chemistry of these tapes provides for good thermal stability of the base polymer.
The thermally conductive tapes are on a silicone treated polyester release liner for ease of handling and die-cutting. The tapes offer both good thermal conductivity and good electrical insulation properties. Best adhesion results when used with high surface energy substrates.