Self-Laminating Vinyl Labels for Thermal Transfer Printers, Brady
Supplier: Brady Worldwide
30550
76242-074EA 167.97 USD
76242-074 76242-076 76242-078 76244-724 76245-016 76247-504 76247-742 76248-974
Self-Laminating Vinyl Labels for Thermal Transfer Printers, Brady
Labels
Self-laminating vinyl material provides a protective layer over text for excellent abrasion, dirt, oils, solvents, water and excess handling resistance.
  • Service temperature of −40 to 158 °F (−40 to 70 °C)
  • Ideal for use in cable and wire labeling, voice-data identification, and panel marking
  • Printable white zone with self-lamination prints on a white background with translucent over laminating area to protect text
Permanent acrylic tape adheres well to surfaces that are smooth, rough or powder coated, and to irregular surfaces or surfaces with low surface energy.
Resistant to chemical, humidity and weather.
Certifications: RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC; UL approved.