Conductive Caps, CEC Series, Caplugs
Supplier: Caplugs
CEC-14 CEC-16 CEC-12 CEC-05 CEC-08 CEC-10 CEC-22 CEC-24 CEC-26 CEC-04
76244-274PK 423.27 USD
76244-274 76244-282 76244-996 76246-390 76246-392 76246-394 76246-396 76246-398 76246-400 76277-282
Conductive Caps, CEC Series, Caplugs
Closures Safety Caps and Safety Waste Caps
CEC series conductive caps are for all common sizes of threaded electrical and electronic connectors. Also suitable for protecting non-threaded bayonet type connectors. In addition to dissipating electrostatic charges, they provide an electrical shield over the end of the connector.
  • Conductive polyethylene copolymer