Saturix White Creped Quarter-Fold Wipes
Supplier: FG CLEAN WIPES TE
799CBDB00
76244-914CS 470.22 USD
76244-914
Saturix White Creped Quarter-Fold Wipes
Wipes Disposable Wipes and Towels Wipes for General Applications
Economical, non-woven, polyester/cellulose wipes are perfect for a variety of everyday and specialized tasks, especially in support work areas. These highly absorbent wipes are designed for less critical applications.
  • Hydroentangled blend of 55% cellulose and 45% polyester (64 g/m²)
  • Creped surface is more effective in picking up particulates and contaminants
  • Strong, absorbent, and versatile wiping and spill control
  • Economical and efficient industrial wipe
The creped surface of the wipe makes it more effective in particulate pick-up and cleaning. These highly absorbent wipes are able to absorb five times (5×) their own weight, making them perfect for a variety of cleaning applications.