Tape, Industrial, Uline
Supplier: ULINE
S-423
76244-898CS 331.25 USD
76244-898
Tape, Industrial, Uline
Tapes
This tape has strong solvent acrylic adhesive sticks to virtually any surface.
  • Stands up to heat, humidity and cold
  • Application range is 0 to 140 °F
  • Long shelf life