Halogen Bulbs, Ushio America
Supplier: USHIO AMERICA ATT. CUST. SERV. TE
5000506
76244-828EA 1060.76 USD
76244-828
Halogen Bulbs, Ushio America
Lights UV Lights, Lamps and Bulbs
Used in fluorescent microscope system, spot UV curing, light guid application.
  • Enhanced electrodes designed for precise positioning and hight arc stability
  • Improved manufacturing process enable exended life