Chicago Latrobe® Heavy-Duty Screw Length Drills, Cutting Tool Supply
Supplier: CUTTING TOOL SUPPLY TE
49202
76244-564EA 4.8 USD
76244-564
Chicago Latrobe® Heavy-Duty Screw Length Drills, Cutting Tool Supply
Tool Sets
Heavy-duty construction for drilling in tough, high-tensile, and work-hardening materials under extreme operating conditions.
  • 135° P3 split point is self-centering for reduced thrust and easier penetration
  • Manufactured from premium cobalt high-speed steel
  • Increasing hot hardness and extending drill wear life
Black oxide surface finish increases wear resistance and adds lubricity, improving chip flow.