Chicago Latrobe® Screw Machine Length Drills, Cutting Tool Supply
Supplier: CUTTING TOOL SUPPLY TE
48705 48730 48707 48721 48508 48529 48512 48530 48701 48702 48708 48715 48716 48728 48731 48740 48743 48751 48757 48760 48519 48521 48522 48524 48526 48527 48528 48531 48532 48704 48710 48717 48724 48734 48505
76244-450EA 5.86 USD
76244-450 76244-452 76244-454 76244-456 76244-458 76244-460 76244-462 76244-464 76244-526 76244-528 76244-530 76244-532 76244-534 76244-536 76244-538 76244-544 76244-548 76244-554 76244-556 76244-558 76244-566 76244-570 76244-574 76244-582 76244-584 76244-586 76244-588 76244-590 76244-592 76254-276 76254-278 76254-280 76254-288 76254-294 76254-304
Chicago Latrobe® Screw Machine Length Drills, Cutting Tool Supply
Tool Sets
These general-purpose screw machine length drills are designed for drilling a wide variety of materials on different range of operating conditions.
  • Made of premium high-speed steel
  • 118°-point style
  • Extra rigid for portable drilling
  • Available in fractional, letter and wire gage size
  • Style 157 is either of bright finish or black oxide coated
The shorter flute and overall length provide superior rigidity to minimize deflection when using heavy fee rates. Bright finish with polished flutes enhances chip ejection especially for aluminum and other non-ferrous materials.