Melin® Single Flute Countersinks, Cutting Tool Supply
Supplier: CUTTING TOOL SUPPLY TE
18001 18011 19200 18003 18002
76244-442EA 45.25 USD
76244-442 76244-444 76244-446 76244-448 76244-678
Melin® Single Flute Countersinks, Cutting Tool Supply
Tool Sets
The radial relief feature of this countersinks ensures smooth, chatter-free cutting without overfeeding and makes the tool self-centering.
  • Excellent countersink for use in hand held applications
Black oxide surface treatment enhances tool life and prevents galling.