Hexacon Spudger Soldering Tools, Jensen Tools
Supplier: TESTEQUITY LLC TE
MA-800G
76244-432EA 15.7 USD
76244-432
Hexacon Spudger Soldering Tools, Jensen Tools
Soldering Supplies Soldering Stations
  • ESD safe, static-dissipative fiberglass polyamide material
  • Electrically and thermally non-conductive
  • Does not scratch printed wiring boards