Metcal Advanced Soldering and Rework Hand-pieces, OK International
Supplier: OK INTERNATIONAL TE
MX-H1-AV
76244-420EA 509.6 USD
76244-420
Metcal Advanced Soldering and Rework Hand-pieces, OK International
Soldering Supplies Soldering Irons
Soldering and rework cartridge hand-piece for the MX-5000 and MX-5200 series systems.
  • Ergonomic and cool-to-the-touch feel
  • Replaceable grip
  • Hand-piece cable length: 178 cm (70")
General purpose tool for single task soldering, touch-up soldering, Surface Mount Device (SMD) rework, and pad clean-up.
Compatible with STTC and SMTC cartridges, MX-W1AV TipSaver workstand.