Wetordry™ Abrasive Sheet, 3M
Supplier: 3M
7000118294
76244-372CS 1021.99 USD
76244-372
Wetordry™ Abrasive Sheet, 3M
Abrasives
Wetordry™ Abrasive Sheet 413Q is an abrasive sanding paper constructed on a light, weight paper backing and features a waterproof resin bonding to help prevent heat buildup.
  • Silicon carbide mineral provides an aggressive cut and a consistent, fine finish
  • Waterproof material allows for easy debris removal
  • Can be easily cut and folded to fit any hand tool or work piece crevice
The silicon carbide abrasive and waterproof backing make this paper an ideal product for sanding applications such as paint prep, sanding, sealer sanding, and solid surface finishing.