Statfree® Document Holder, Desco Industries
Supplier: Desco
07454
76244-042PK 147.7 USD
76244-042
Statfree® Document Holder, Desco Industries
Document Holders
Dissipative Statfree® document holders are designed for holding paperwork in ESD protected areas.
  • Heavyweight of 6 mil, top-loading document holder
  • Non-stick, amine-free, coextruded polyolefin material
Designed for holding paperwork (8.5×11") in ESD protected areas. Triboelectric charge generation is negligible. Non-stick, amine-free, coextruded polyolefin material allows for easy loading of documents, and will not introduce containments to the work area. Lead-free RoHS compliant.
Certifications: Tested per EIA-541; will not expose ESD susceptible items to harmful ESD charges.