Carbofib 3-CF Tweezers, Aven Tools
Supplier: Aven Tools
18702
76243-982EA 120.1 USD
76243-982
Carbofib 3-CF Tweezers, Aven Tools
Forceps
These Swiss made, ESD safe tweezers feature excellent wear and abrasion resistance.
  • High rigidity, excellent tensile and flexural strength
  • Good chemical resistance to oils, grease, fuels, and nonpolar solvents
Ideal for handling sensitive materials such as electronic components, micro-mechanical parts, glass and ceramic substrates.