Technik™ Precision Round Nose Pliers, Aven
Supplier: Aven Tools
10305
76243-980EA 43.56 USD
76243-980
Technik™ Precision Round Nose Pliers, Aven
Pliers
Pliers feature dual leaf springs and box joint construction for comfortable action.
  • Induction hardened cutting edges
  • ESD-safe grips
  • Ideal for forming or coiling fine wires
Made of mirror polished stainless steel which provides superior rust protection compared with standard carbon steel pliers.