Temp-Plate® Standard Four Position Recorder, Wahl Instruments
Supplier: WAHL INSTRUMENTS INC. TE
240-082C
76243-788BX 95.29 USD
76243-788
Temp-Plate® Standard Four Position Recorder, Wahl Instruments
Chart Recorders Recorders
Temp-Plate® standard four position recorder's careful selection of temperature points will allow detection and recording of most overheat occurrences.
  • The unit of measure is box
  • Good for general purpose maintenance
Temp-Plate® cover material is made of Mylar (clear) for ranges 90/350 °F (32/176 °C) and Kapton (amber) for 350/500 °F (176/260 °C).
Delivery information: Temp-Plate® are shipped in reclosable storage boxes of ten labels.