Objectives for LX Microscopes by Unitron
Supplier: LX Microscopes by UNITRON
23750
76243-762EA 402.43 USD
76243-762
Objectives for LX Microscopes by Unitron
Microscope Objectives
These lens are recommended only for use with LX Microscopes by Unitron System's 273 and 373.
  • Field of view is 65.7 to 10.2 mm
  • Total magnification ranges from 3.5 to 22.5×