LED Ring Light, Unitron Ltd
Supplier: LX Microscopes by UNITRON
LED3000
76243-760EA 920.72 USD
76243-760
LED Ring Light, Unitron Ltd
Lights LED Lights, Lamps and Bulbs
LED Ring light with polarizing filter fits most microscopes. It maximizes light output and distribution while preventing harmful glare and reflections. The 40 LED display makes this ring light ideal for the inspection of surface cracks and defects as well as for lead-free inspection.
  • ESD-Safe
  • Five-year warranty
  • 40 individual pure-white LEDs which consume 9 W of power
  • 50000 hour lumen maintenance (lamp life)
  • Up to 3600 footcandles at 4.5" working distance
  • Self-contained on/off and dimming controls, no need for an external controller
Pure-white LED light eases the examination of welds, solder joints, flux residues and hairline stress cracks. Comes complete with a removable/replaceable diffuser cover which allows for easy cleaning and LED array protection from damage and contamination.