3M™ Heavy Duty Duct Tape, 3M
Supplier: 3M
7000028933
76243-706CS 1236.99 USD
76243-706
3M™ Heavy Duty Duct Tape, 3M
Tapes
3M™ Duct tape is composed of waterproof polyethylene film over cloth scrim backing with a rubber adhesive used for demanding duct tape applications.
  • Offers easy tear and conformability
  • Low tack roll edges so edges stay cleaner for less surface contamination and less waste
  • Individual roll wrap that preserves integrity of the roll for less waste
  • Polyethylene coated cloth scrim that resists moisture to prolong tape bond
  • Good tensile strength for tough applications
  • Rubber adhesive that adheres aggressively to most surfaces for good holding power and a tight seal
Features polyethylene film over cloth scrim backing.
Certifications: ASTM D3652.