Zinc Plated Steel Handle Applicator Brush, Gordon Brush
Supplier: Gordon Brush
1HH
76243-700EA 46.25 USD
76243-700
Zinc Plated Steel Handle Applicator Brush, Gordon Brush
Brushes
This zinc plated steel handle applicator brush is an inexpensive utility duster/cleaner that is an excellent replacement for disposable acid brushes that quickly shed their fibers.
  • Anti-static
  • Adhesive-free to prevent leaching and contamination of surface areas
  • Brush fibers are secured using a zinc plated steel handle, substituting adhesives with 5 tons of mechanical clamping