Glass Cloth Electrical Tape 69, 3M
Supplier: 3M
7000145501
76243-652CS 1367.95 USD
76243-652
Glass Cloth Electrical Tape 69, 3M
Tapes
Glass Cloth Electrical Tape 69 is a woven glass cloth tape with a silicone pressure-sensitive adhesive.
  • Printable
  • UL Recognized 200 °C