Round Nose Pliers, Swanstrom Tools USA
Supplier: SWANSTROM TOOLS USA TE
S236E-TP
76243-552EA 128.76 USD
76243-552
Round Nose Pliers, Swanstrom Tools USA
Pliers
These ergonomic style round nose pliers are ESD safe (static dissipative) which protects circuit components.
  • Does not insulate
  • Do not use on live circuits
These superior quality tools are forged from high chromium, high carbon alloy steel.