IBC™ Cleaning Tools, Standard Series, US Conec LTD
Supplier: US CONEC LTD. CUSTOMER SERVICE TE
9393 12910
76243-500EA 215.96 USD
76243-500 76248-870
IBC™ Cleaning Tools, Standard Series, US Conec LTD
Safety Glasses
The IBC™ Cleaners are mechanical cleaning tools designed to clean connectors residing in an adapter or faceplate and unmated connectors.
  • Crush resistant to over 250 N
  • Impact resistant to survive drops over 1.5 M
  • Simple pushing motion to engage tool
The IBC™ Brand cleaning tools use a novel dry cleaning strand to gently sweep and lift away dust and residues from the connector end face. Dry cleaning strand eliminates the need for solvents to be effective.