3M™ VHB™ Tape 4956, 3M
Supplier: 3M
7000148374
76243-416CS 5373.73 USD
76243-416
3M™ VHB™ Tape 4956, 3M
Tapes
This is a nylon film tape with a non-silicone rubber adhesive. Non-silicone adhesive allows for clean removal, great for parts requiring additional painting or bonding.
  • Features excellent performance in high temperature bake cycles (400 °F/204 °C)
  • Strong backing removes quickly for simple application
  • Nylon backing offers toughness and high elongation
  • Strong and very thick
  • Helps remove flashing after composite bonding
This tape is used where thicker caliper nylon tape is required and in advanced composite and metal bond masking.