Aluminum Handle Scratch Brushes, Gordon Brush
Supplier: Gordon Brush
33HHA
76243-290EA 65.34 USD
76243-290 76249-310
Aluminum Handle Scratch Brushes, Gordon Brush
Brushes
Brushes with anti-static horsehair bristle and a hand-laced aluminum handle.
  • Horsehair fill material is hand-laced with stainless steel wire into its handle for maximum fiber retention
  • With two or three rows of bristles
  • Anti-static
Ideal for detail cleaning on items such as printed circuit boards and SMT assemblies, or for use as fluid applicators. Suitable for use in an ESD environment provided the brush remains saturated in liquid. It is recommended that the brush be tested on the required application to confirm ESD compatibilty.