Anti-Shock Shear Cutters, Excelta
Supplier: Excelta
530E-USS-020
76243-266EA 497.48 USD
76243-266 76248-640 76253-040
Anti-Shock Shear Cutters, Excelta
Cutters
The 'Stand-Off' of these cutters keeps the cutting blade from cutting into the solder connections.
  • The grips are static dissipative
These cutters reduce shock on delicate components and solder connections.