FR-830 Preheater, American Hakko Products
Supplier: AMERICAN HAKKO PRODUCTS INC TE
FR830-02
76243-208EA 1212.47 USD
76243-208
FR-830 Preheater, American Hakko Products
Tool Sets
The FR-830 is a compact and low cost solution to preheating printed circuit boards. The design features on the FR-830 deliver consistent heat to the bottom of printed circuit boards and is particularly beneficial when soldering lead-free assemblies.
  • Compact design
  • Fast heating
  • Excellent thermal stability
  • ESD-safe by design
The integrated self-resetting overheat protection, high pressure fan, and convex outlet reduces heat buildup and prevents blockage of the outlet, and the angled heater tube protects the heating element from damage caused by debris that may accidentally drop inside the heater opening.