VERSATOUCH® 88-356 Glove, Magid
Supplier: Magid Glove
88-356-7
76243-192PK 54.59 USD
76243-192
VERSATOUCH® 88-356 Glove, Magid
Gloves
Lightweight, unlined natural rubber glove.
  • Food approved
  • Light duty glove with atterned grip
  • Good chemical, snag, puncture and abrasion resistance
Blue coating widely preferred by the poultry and food processing industry, with 17 mil for good tactile sensitivity and dexterity.
Certifications: Compliant with FDA food handling requirements.