Paper Tape 200, 3M
Supplier: 3M
7000144751
76243-174CS 114.02 USD
76243-174
Paper Tape 200, 3M
Tapes
Paper Tape 200 is a utility purpose paper tape for holding, bundling, sealing, non-critical masking tape and a vast number of other jobs where a pressure-sensitive tape is needed.
  • Provides good holding power to help resist lifting or curling
  • Uses solvent free manufacturing process
  • Good instant adhesion that sticks to a wide variety of surfaces
The crepe paper provides conformability for easy handling and application. Is easy and tearable, yet resists slivering. Special backsize treatment provides easy to use controlled unwind. It is not recommended for paint masking, but may be used for non-critical applications. It is designed for indoor use only, and should not be subjected to outdoor or UV exposure.