Dispensing Piston, OK International
Supplier: OK INTERNATIONAL TE
93055-WW
76243-094BX 95.67 USD
76243-094
Dispensing Piston, OK International
Fluid Adhesive Dispensers and Pistons
Wiper pistons provide a seal for low to medium viscosity fluids. It is made from polyethylene