Syringe Barrel Pistons, EFD
Supplier: EFD INC.- CUSTOMER SERVICE TE
7015479
76238-210BX 62.02 USD
76238-210 76241-004 76241-734 76241-736 76243-038 76243-854 76245-256 76245-272 76245-716 76247-664 76248-740 76251-460 76252-446
Syringe Barrel Pistons, EFD
Syringe Barrels, Plungers and Pistons
A piston is inserted into the syringe barrel after it has been loaded with fluid to ensure uniform dispensing, prevent dripping, and eliminate waste by wiping barrel walls clean as fluid is dispensed.
  • White SmoothFlow™ wiper pistons are used with most fluids
  • Beige SmoothFlow™ pistons are loose-fitting and used with air-entrapped fluids
  • Red SmoothFlow™ pistons are tight-fitting and used with mechanical dispensers
  • Orange Flat-walled pistons have a looser fit to prevent “bouncing” when dispensing stringy, air-entrapped fluids
  • Blue LV Barrier pistons are for cyanoacrylates and very low viscosity fluids