CLEANimage Cleanroom Papers, Purus International
Supplier: VWR International
LCIY-2012C
76242-962CS 326.85 USD
76242-962
CLEANimage Cleanroom Papers, Purus International
Paper
Cleanroom papers for writing, printing and photocopying in cleanroom environments.
  • Dual binding technology
  • Excellent printability and heat toner adhesion
  • Double sided web cleaned prior to sheeting
  • High opacity and durability
Pure image Premier Paper produced in Class 10 (ISO 4) cleanroom. Suitable for use in Class 10 to Class 1000 (ISO class 4 to Class 6) cleanroom.