Performus™ Precision Fluid Dispensers, EFD
Supplier: EFD INC.- CUSTOMER SERVICE TE
7012330
76242-896EA 3738 USD
76242-896 76247-818
Performus™ Precision Fluid Dispensers, EFD
Fluid Adhesive Dispensers and Pistons
EFD’s Performus™ dispensers increase throughput, improve yields, and reduce production costs through controlled application of adhesives, lubricants and other assembly fluids.
  • Handles all fluids
  • Vacuum control keeps thin fluids from dripping
  • Features electric foot pedal
Certifications: CE, CSA, RoHS, WEEE and RoHS Compliant.