Sticklers Benchtop CleanWipes™ for Fiber Optics, Jensen Tools
Supplier: TESTEQUITY LLC TE
MCC-WFW
76242-880EA 61.33 USD
76242-880
Sticklers Benchtop CleanWipes™ for Fiber Optics, Jensen Tools
Wipes Disposable Wipes and Towels Lens Papers
CleanWipes™ 90 benchtop wipes are high quality, lint-free fiber optic wipes engineered for use on bare fiber before splicing, and also for cleaning fiber optic jumpers.
  • Optical-frade fabric in convenient mini-tub
  • Made without glues, bleach or cellulose
  • Stronger and less linting than cellulose wipes
  • Superior results when cleaning bare fiber
  • Affordable on a cost-per-cleaning basis
They are exactly the right size and the right material to make wet-dry cleaning simple and cost-effective.
Packaging: Each tub contains 90 perforated pop-up 4×2" lint-free wipes.