Scotch® Insulation Tape 22, Vinyl, 3M
Supplier: 3M
7000042539
76242-674CS 1388.52 USD
76242-674
Scotch® Insulation Tape 22, Vinyl, 3M
Tapes
Scotch® vinyl insulation tape 22 is a premium-grade, heavy-duty, vinyl electrical tape designed for insulation applications that require greater mechanical strength and abrasion resistance.
  • Provides moisture-resistant electrical and mechanical protection
  • Provides high resistance to UV rays, abrasion, moisture, alkalis, acids, corrosion and varying weather conditions
  • Made of black durable PVC
  • Withstands a temperature range of –10 to 80 °C (15 to 176 °F)
It is suitable for both indoor and outdoor applications. The tape is compatible with solid dielectric cable insulations, synthetic cable insulations, jackets and splicing compounds.
Certifications: UL Listed, ASTM Approved, VDE, CSA Certified and RoHS 2011/65/EU Compliant.