Ground Cord for Floor Mat, Menda
Supplier: Desco
09816
76242-576EA 17.22 USD
76242-576
Ground Cord for Floor Mat, Menda
Grounders Grounding Wires
Ground cord terminated with a banana plug, easily snaps to mat socket, flush to mat surface.
Stud has threaded hole; 6 to 32×1/4" flat head screw and countersunk washer are provided allows ground cord to be optionally bolted mat; keeps cord from disconnecting; ensures proper grounding.
Certifications: Meets grounding standard ANSI/ESD S6.1.