Electrical Tape, Polyimide Film, 3M
Supplier: 3M
7000132698
76242-356CS 1887.43 USD
76242-356
Electrical Tape, Polyimide Film, 3M
Tapes
Polyimide film electrical tape is amber with a 3-mil polyimide film backing with an acrylic adhesive system for exceptional heat and solvent resistance and may be applied at very low temperatures.
  • This tape can be imprinted