Copper Foil Shielding Tapes, 3M
Supplier: 3M
7100081931
76242-354CS 2562.58 USD
76242-354
Copper Foil Shielding Tapes, 3M
Tapes
Dead soft 3.5-mil copper foil backing for good shielding effectiveness, soldering and converting.
  • Removable liner on both sides makes it easy to handle and die cut complex shapes
  • Double-coated conductive adhesive makes an excellent conductive gasket
Conductive particles in the adhesive on both sides provides low contact resistance between substrates and the backing to drain static charge.