Angle End Cutters, Swanstrom Tools USA
Supplier: SWANSTROM TOOLS USA TE
S225E-TP
76242-322EA 279.77 USD
76242-322
Angle End Cutters, Swanstrom Tools USA
Cutters
These ergonomic style angle end cutters are ESD safe (static dissipative) which protects circuit components.
  • Does not insulate
  • Do not use on live circuits
These superior quality tools are forged from high chromium, high carbon alloy steel.