HyFlex® 11-801 Gloves, Ansell
Supplier: Ansell Healthcare
205674
76242-264CS 1318.64 USD
76242-264
HyFlex® 11-801 Gloves, Ansell
Gloves
HyFlex® 11-801 glove, the original nitrile foam gloves, is better than ever is highly versatile solution for precision assembly and general handling, it provides the perfect balance between comfort, dexterity and protection.
  • Darker liner is well-suited for slightly oily or dirty environments helping to improve wear life
  • No transfer of silicone contaminants to metal parts prior to painting
Advanced Knitting Technology allows increased stretch and flexibility in high-stress areas: the hand operates more freely, improving comfort and reducing hand fatigue.