Anti-Static Poly Bag, Uline
Supplier: ULINE
S-6583
76242-240BX 89.48 USD
76242-240
Anti-Static Poly Bag, Uline
Bags Sample Bags
This bag safeguards electronic components from static build-up.
  • Anti-static
  • Non-corrosive
  • Made of virgin polyethylene film
Heat sealable with impulse sealers.