Small Upright Applicator Brush, Gordon Brush
Supplier: Gordon Brush
WA12CK
76241-974EA 15.06 USD
76241-974
Small Upright Applicator Brush, Gordon Brush
Brushes
Small upright applicator brush with anti-static hog bristle and a static-neutral wooden handle.
  • Heavy-duty scrubbing and duster brush that works well for getting into tight areas
  • Also known as a typewriter or keyboard brush
  • 2 x 6 row configuration, with a 1/2" trim, and a brush face of 7/8" x 3/8"
Ideal for detail cleaning, such as printed circuit boards and SMT assemblies, or for use as fluid applicators.