Syringe Adapters, X-Series, Jensen Tools
Supplier: JENSEN GLOBAL INC
JG30A-X6 JG5A-X6 JG10A-X6
76241-818EA 101.23 USD
76241-818 76244-012 76244-016
Syringe Adapters, X-Series, Jensen Tools
Syringe Barrels, Plungers and Pistons
Syringe adapters are specifically designed with an o-ring seal to fit all Jensen Global air syringes. It uses 1/4" air line secured with a clamp.