Cut Off Wheel, Dremel
Supplier: DREMEL SERVICES INC. TE
DR-409
76241-456EA 26.9 USD
76241-456
Cut Off Wheel, Dremel
Cutters
Cut-off wheel make it easy to cut or slot bolts, screws, sheet metal, thin wood and plastic.
  • Cuts only along its edge
  • Do not attempt to sand or cut curved holes with them
For slicing and cutting metal, wood and plastic.