Cartridge Blade 10 S1, JBC
Supplier: JBC TOOLS USA INC. TE
C245914
76241-410EA 269.28 USD
76241-410
Cartridge Blade 10 S1, JBC
Tool Sets
This cartridge has a 10 mm wide blade tip.
C245 cartridge range is the widest, with many shapes, sizes and special models. They work with T245 handpieces.
They are appropriate for general soldering purposes and they are perfect for both SMD components or for applications with high power requirements such as ground plane soldering.