Transverse End Cutters, Swanstrom Tools USA
Supplier: SWANSTROM TOOLS USA TE
S233E-TP
76241-346EA 176.77 USD
76241-346
Transverse End Cutters, Swanstrom Tools USA
Cutters
These ergonomic style transverse end cutters are ESD safe (static dissipative) which protects circuit components.
  • Does not insulate
  • Do not use on live circuits
These superior quality tools are forged from high chromium, high carbon alloy steel.