SIRUS® Surgical Gowns, Medline Industries
Supplier: Medline
DYNJP2209
76241-306CS 565.24 USD
76241-306
SIRUS® Surgical Gowns, Medline Industries
Disposable Garments
Sirus surgical gowns offer a high level of comfort. Dependable AAMI level 3 and AAMI level 4 protection comes from five fabric layers, three densely-packed meltblown layers sandwiched between two strong spunbound outer layers, for an excellent barrier against fluids and pathogens.
  • Roominess and a larger sweep make movement easier