PurSwab® Chemical Resistant Paddle Shaped Foam Swab, Hardwood Products Company
Supplier: PURITAN MEDICAL PRODUCTS COMPANY I
1876-PF CR
76241-190CS 793.6 USD
76241-190
PurSwab® Chemical Resistant Paddle Shaped Foam Swab, Hardwood Products Company
Swabs Swabs for Clinical Use
Chemical resistant foam paddle tipped applicator with plastic handle. Can be used with acetone.
  • Slightly textured foam tip for light abrasion
  • Tip will not swell or break apart with most solvents
General cleaning swab, ideal for removing and applying adhesives and other solutions.