CleanWIPE® Swab, Semi-Flexible Mini Spear Tip, Foamtec
Supplier: FOAMTEC INTERNATIONAL, LLC.
HT1049-500
76241-188CS 3802.28 USD
76241-188
CleanWIPE® Swab, Semi-Flexible Mini Spear Tip, Foamtec
Swabs Swabs for Cleaning
CleanWIPE Swabs are designed for the most demanding cleanroom precision cleaning operations. 1000 Series Swabs employ the CleanWIPE® reticulated 100 ppi foam to provide soft, absorbent cleaning performance. The CleanWIPE® swabs are available in a variety of head shapes and sizes enabling precision cleaning of optical, disk drive, and class III medical device components. Swab handle and tip materials are traceable back to raw materials.
  • Swab tip is constructed from medical grade open-cell PU foam to enable efficient removal of silicones, adhesives and epoxies
  • Super fine seal eliminates risk of scratching delicate optical assemblies
  • Silicone and adhesive free to ensure streak free cleaning when wetted with solvents such as IPA
  • High purity formulation and multistage washing process ensure a swab free of Amines, Sulphur and Chloride to ensure compatibility with curing of platinum cured silicones, adhesive and epoxies
Certifications: Swab manufacturing, cleaning and packaging is performed in an ISO 13485, ISO 5 cleanroom to ensure maximum cleanliness and lot to lot uniformity.